3-Bromo-2-chlorofuran ≥95%

Supplier: AOBChem USA

Warning

46741-1G 46741-5G 46741-500MG
77639-020EA 676 USD
77639-020 77659-520 77647-024
3-Bromo-2-chlorofuran ≥95%
3-Bromo-2-chlorofuran

Precaución: For Research Use Only

Formula: C₄H₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD28667690
CAS Number: 31491-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR