1-Bromo-2-iodo-4-(trifluoromethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

46738-5G 46738-25G
77635-044EA 178 USD
77635-044 77659-518
1-Bromo-2-iodo-4-(trifluoromethoxy)benzene ≥97%
1-Bromo-2-iodo-4-(trifluoromethoxy)benzene

Precaución: For Research Use Only

Formula: C₇H₃BrF₃IO
Storage Temperature: Ambient
MDL Number: MFCD12025452
CAS Number: 1121586-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR