2-Methoxy-1-(4-phenoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

467297-1G 467297-5G 467297-500MG
77639-012EA 560 USD
77639-012 77659-514 77647-012
2-Methoxy-1-(4-phenoxyphenyl)ethanone ≥95%
2-Methoxy-1-(4-phenoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 242.27 g/mol
MW: C₁₅H₁₄O₃
Storage Temperature: Ambient
CAS Number: 1216293-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR