2-Chloro-1-(4-phenoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

467296-5G 467296-25G 467296-1G 467296-500MG
77633-254EA 639 USD
77633-254 77655-942 77643-044 77647-010
2-Chloro-1-(4-phenoxyphenyl)ethanone ≥95%
2-Chloro-1-(4-phenoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 246.689 g/mol
MW: C₁₄H₁₁ClO₂
Storage Temperature: Ambient
CAS Number: 13075-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR