(4-Chloro-2-methoxyphenyl)(cyclopropyl)methanone ≥95%

Supplier: AOBChem USA

Warning

467294-5G 467294-1G 467294-500MG
77629-580EA 1484.23 USD
77629-580 77643-042 77651-016
(4-Chloro-2-methoxyphenyl)(cyclopropyl)methanone ≥95%
(4-Chloro-2-methoxyphenyl)(cyclopropyl)methanone

Precaución: For Research Use Only

Formula: C₁₁H₁₁ClO₂
Storage Temperature: Ambient
CAS Number: 1600973-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR