2-(7-Methoxynaphthalen-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

467258-5G 467258-1G 467258-25G 467258-500MG
77629-574EA 1738.64 USD
77629-574 77639-006 77659-508 77647-002
2-(7-Methoxynaphthalen-2-yl)propan-2-ol ≥95%
2-(7-Methoxynaphthalen-2-yl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₄H₁₆O₂
Storage Temperature: Ambient
CAS Number: 3031798-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR