1-(4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

467241-5G 467241-25G 467241-1G 467241-500MG
77629-572EA 1484.23 USD
77629-572 77655-938 77643-036 77651-014
1-(4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 339.004 g/mol
MW: C₉H₂BrF₇O
Storage Temperature: Ambient
CAS Number: 3005483-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR