2-(4-Chloro-2-(cyclopropylmethoxy)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

467213-5G 467213-25G 467213-1G 467213-500MG
77629-570EA 1484.23 USD
77629-570 77655-936 77643-032 77651-010
2-(4-Chloro-2-(cyclopropylmethoxy)phenyl)propan-2-ol ≥95%
2-(4-Chloro-2-(cyclopropylmethoxy)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 240.726 g/mol
MW: C₁₃H₁₇ClO₂
Storage Temperature: Ambient
CAS Number: 3031788-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR