Tert-butyl 2-(benzyloxy)-3-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

467190-5G 467190-25G 467190-1G 467190-500MG
77629-566EA 1768.51 USD
77629-566 77635-884 77643-028 77647-000
Tert-butyl 2-(benzyloxy)-3-methylbenzoate ≥95%
Tert-butyl 2-(benzyloxy)-3-methylbenzoate

Precaución: For Research Use Only

Formula: 298.376 g/mol
MW: C₁₉H₂₂O₃
Storage Temperature: Ambient
CAS Number: 3031798-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR