Ethyl 2-(2-(benzyloxy)-3-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

467189-5G 467189-25G 467189-1G 467189-500MG
77629-564EA 1527.49 USD
77629-564 77635-882 77643-026 77646-998
Ethyl 2-(2-(benzyloxy)-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2-(benzyloxy)-3-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 298.333 g/mol
MW: C₁₈H₁₈O₄
Storage Temperature: Ambient
CAS Number: 3031799-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR