1-(4-(Cyclopentyloxy)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

467184-5G 467184-25G 467184-1G 467184-500MG
77629-562EA 1484.23 USD
77629-562 77655-934 77643-024 77646-996
1-(4-(Cyclopentyloxy)phenyl)propan-1-one ≥95%
1-(4-(Cyclopentyloxy)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 218.292 g/mol
MW: C₁₄H₁₈O₂
Storage Temperature: Ambient
CAS Number: 1019367-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR