2,2,3,3,4,4,4-Heptafluorobutyl methanesulfonate ≥95%

Supplier: AOBChem USA

Warning

467072-5G 467072-25G 467072-1G 467072-500MG
77633-250EA 1338 USD
77633-250 77655-932 77643-022 77646-994
2,2,3,3,4,4,4-Heptafluorobutyl methanesulfonate ≥95%
2,2,3,3,4,4,4-Heptafluorobutyl methanesulphonate

Precaución: For Research Use Only

Formula: 278.145 g/mol
MW: C₅H₅F₇O₃S
Storage Temperature: Ambient
CAS Number: 755-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR