2-bromo-3-fluoro-5-methoxyphenol ≥95%

Supplier: AOBChem USA

Warning

46619-1G 46619-5G 46619-500MG
77639-004EA 635 USD
77639-004 77659-502 77651-006
2-bromo-3-fluoro-5-methoxyphenol ≥95%
2-bromo-3-fluoro-5-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₆BrFO₂
MW: 221.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738848
CAS Number: 1785528-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR