2-Methyl-4-(3-methylphenyl)-3-butyn-2-ol ≥95%

Supplier: AOBChem USA

Warning

465498-5G 465498-1G 465498-500MG
77659-500EA 2001 USD
77659-500 77643-018 77646-990
2-Methyl-4-(3-methylphenyl)-3-butyn-2-ol ≥95%
2-Methyl-4-(3-methylphenyl)-3-butyn-2-ol

Precaución: For Research Use Only

Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Storage Temperature: Ambient
CAS Number: 63079-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR