Methyl 3-amino-5,6-dibromo-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

465367-5G 465367-25G 465367-1G 465367-500MG
77629-558EA 1484.23 USD
77629-558 77635-880 77643-016 77651-004
Methyl 3-amino-5,6-dibromo-2-methylbenzoate ≥97%
Methyl 3-amino-5,6-dibromo-2-methylbenzoate

Precaución: For Research Use Only

Formula: 322.9812 g/mol
MW: C₉H₉Br₂NO₂
Storage Temperature: Ambient
CAS Number: 3031812-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR