Methyl 2,3-dibromo-6-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

465358-5G 465358-25G 465358-1G 465358-500MG
77629-556EA 1484.23 USD
77629-556 77635-878 77643-014 77651-002
Methyl 2,3-dibromo-6-methylbenzoate ≥97%
Methyl 2,3-dibromo-6-methylbenzoate

Precaución: For Research Use Only

Formula: 307.9666 g/mol
MW: C₉H₈Br₂O₂
Storage Temperature: Ambient
CAS Number: 2383186-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR