1-(2,4-Diethylphenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

465323-5G 465323-1G 465323-25G 465323-500MG
77629-554EA 1484.23 USD
77629-554 77639-002 77659-498 77651-000
1-(2,4-Diethylphenyl)cyclobutanol ≥95%
1-(2,4-Diethylphenyl)cyclobutanol

Precaución: For Research Use Only

Formula: 204.308 g/mol
MW: C₁₄H₂₀O
Storage Temperature: Ambient
CAS Number: 3031799-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR