(4-(Cyclopentyloxy)phenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

465221-5G 465221-25G 465221-1G 465221-500MG
77633-248EA 1441 USD
77633-248 77655-930 77643-012 77646-986
(4-(Cyclopentyloxy)phenyl)(ethyl)sulfane ≥95%
(4-(Cyclopentyloxy)phenyl)(ethyl)sulphane

Precaución: For Research Use Only

Formula: 222.346 g/mol
MW: C₁₃H₁₈OS
Storage Temperature: Ambient
CAS Number: 3031799-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR