Cyclohexyl(4-fluoro-2-methylphenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

465079-5G 465079-1G 465079-500MG
77659-494EA 2001 USD
77659-494 77643-010 77646-982
Cyclohexyl(4-fluoro-2-methylphenyl)methanone ≥97%
Cyclohexyl(4-fluoro-2-methylphenyl)methanone

Precaución: For Research Use Only

Formula: 220.283 g/mol
MW: C₁₄H₁₇FO
Storage Temperature: Ambient
CAS Number: 1338973-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR