2,2,2-Trifluoro-1-(2-fluoro-3-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

465061-5G 465061-500MG 465061-25G 465061-1G
77629-546EA 1484.23 USD
77629-546 77652-858 77659-492 77643-008
2,2,2-Trifluoro-1-(2-fluoro-3-propoxyphenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(2-fluoro-3-propoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 250.19 g/mol
MW: C₁₁H₁₀F₄O₂
Storage Temperature: Ambient
CAS Number: 3037682-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR