2-(2-Fluoro-3-propoxyphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

465060-5G 465060-25G 465060-1G 465060-500MG
77629-544EA 1484.23 USD
77629-544 77655-928 77643-006 77646-980
2-(2-Fluoro-3-propoxyphenyl)propan-2-ol ≥95%
2-(2-Fluoro-3-propoxyphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 212.261 g/mol
MW: C₁₂H₁₇FO₂
Storage Temperature: Ambient
CAS Number: 3037705-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR