1-(2-Fluoro-3-propoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

465058-5G 465058-25G 465058-1G 465058-500MG
77629-542EA 1484.23 USD
77629-542 77655-926 77643-004 77646-978
1-(2-Fluoro-3-propoxyphenyl)ethanone ≥95%
1-(2-Fluoro-3-propoxyphenyl)ethanone

Precaución: For Research Use Only

Formula: 196.218 g/mol
MW: C₁₁H₁₃FO₂
Storage Temperature: Ambient
CAS Number: 3037695-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR