2-Fluoro-1-iodo-3-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

465056-5G 465056-1G 465056-25G 465056-500MG
77629-540EA 1484.23 USD
77629-540 77638-994 77655-922 77650-998
2-Fluoro-1-iodo-3-propoxybenzene ≥97%
2-Fluoro-1-iodo-3-propoxybenzene

Precaución: For Research Use Only

Formula: C₉H₁₀FIO
MW: 280.078 g/mol
Storage Temperature: Ambient
CAS Number: 3037695-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR