3-(2-Chloro-4-fluorophenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

465052-5G 465052-1G 465052-25G 465052-500MG
77629-538EA 1484.23 USD
77629-538 77638-992 77655-920 77650-996
3-(2-Chloro-4-fluorophenyl)oxetan-3-ol ≥95%
3-(2-Chloro-4-fluorophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: 202.61 g/mol
MW: C₉H₈ClFO₂
Storage Temperature: Ambient
CAS Number: 3031808-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR