2-(4-Methyl-3-(methylthio)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

465048-1G 465048-5G 465048-500MG
77638-990EA 583 USD
77638-990 77655-918 77650-994
2-(4-Methyl-3-(methylthio)phenyl)propan-2-ol ≥95%
2-(4-Methyl-3-(methylthio)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
CAS Number: 1896341-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR