1-(4-Methyl-3-(methylthio)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

465047-5G 465047-25G 465047-1G 465047-500MG
77629-536EA 1484.23 USD
77629-536 77655-916 77643-000 77650-992
1-(4-Methyl-3-(methylthio)phenyl)propan-1-one ≥95%
1-(4-Methyl-3-(methylthio)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 194.293 g/mol
MW: C₁₁H₁₄OS
Storage Temperature: Ambient
CAS Number: 3031792-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR