(5-Iodo-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

464904-5G 464904-1G 464904-25G 464904-500MG
77629-534EA 1484.23 USD
77629-534 77638-988 77659-490 77646-974
(5-Iodo-2-methylphenyl)(methyl)sulfane ≥97%
(5-Iodo-2-methylphenyl)(methyl)sulphane

Precaución: For Research Use Only

Formula: 264.127 g/mol
MW: C₈H₉IS
Storage Temperature: Ambient
CAS Number: 3031792-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR