Trimethyl(4-methyl-3-(methylthio)phenyl)silane ≥95%

Supplier: AOBChem USA

Warning

464902-5G 464902-25G 464902-1G 464902-500MG
77629-532EA 1484.23 USD
77629-532 77635-874 77638-986 77650-990
Trimethyl(4-methyl-3-(methylthio)phenyl)silane ≥95%
Trimethyl(4-methyl-3-(methylthio)phenyl)silane

Precaución: For Research Use Only

Formula: C₁₁H₁₈SSi
Storage Temperature: Ambient
CAS Number: 3031813-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR