5-Bromo-3-cyclopropyl-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
464849-5G
464849-25G
464849-1G
464849-500MG
77633-244EA
931
USD
77633-244
77655-914
77642-998
77650-988
5-Bromo-3-cyclopropyl-1H-indazole ≥95%
5-Bromo-3-cyclopropyl-1H-indazole
Precaución: For Research Use Only
Formula:
C₁₀H₉BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD18837635 CAS Number: 911305-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...