4-(5-Fluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%

Supplier: AOBChem USA

Warning

464823-5G 464823-1G 464823-500MG 464823-25G
77633-242EA 1381 USD
77633-242 77638-982 77652-856 77659-488
4-(5-Fluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Fluoro-2-methylphenyl)tetrahydro-2H-pyran-4-ol

Precaución: For Research Use Only

Formula: 210.2447 g/mol
MW: C₁₂H₁₅FO₂
Storage Temperature: Ambient
CAS Number: 1339644-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR