2-(5-Fluoro-2-methylphenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

464819-5G 464819-1G 464819-500MG
77655-910EA 2250 USD
77655-910 77642-996 77650-984
2-(5-Fluoro-2-methylphenyl)propan-2-ol ≥95%
2-(5-Fluoro-2-methylphenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 168.2080 g/mol
MW: C₁₀H₁₃FO
Storage Temperature: Ambient
CAS Number: 83208-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR