2-(4-Chloro-3-fluorophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

464758-5G 464758-1G 464758-25G 464758-500MG
77633-238EA 1186 USD
77633-238 77638-978 77655-908 77650-980
2-(4-Chloro-3-fluorophenyl)propan-2-ol ≥95%
2-(4-Chloro-3-fluorophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₉H₁₀ClFO
Storage Temperature: Ambient
CAS Number: 1379358-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR