Cyclohexyl(2,3-difluorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

464730-5G 464730-1G 464730-25G 464730-500MG
77633-234EA 1186 USD
77633-234 77638-976 77655-906 77646-970
Cyclohexyl(2,3-difluorophenyl)methanone ≥95%
Cyclohexyl(2,3-difluorophenyl)methanone

Precaución: For Research Use Only

Formula: 224.246 g/mol
MW: C₁₃H₁₄F₂O
Storage Temperature: Ambient
CAS Number: 1344347-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR