Ethyl 2-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

464723-5G 464723-25G 464723-1G 464723-500MG
77629-528EA 1484.23 USD
77629-528 77635-872 77638-974 77650-978
Ethyl 2-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)-2-oxoacetate ≥97%
Ethyl 2-(2,2-difluorobenzo[d][1,3]dioxol-4-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 258.175 g/mol
MW: C₁₁H₈F₂O₅
Storage Temperature: Ambient
CAS Number: 531508-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR