(6-Chloro-2-fluoropyridin-3-yl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

464721-5G 464721-1G 464721-25G 464721-500MG
77629-524EA 1484.23 USD
77629-524 77638-970 77655-904 77650-976
(6-Chloro-2-fluoropyridin-3-yl)(phenyl)methanol ≥95%
(6-Chloro-2-fluoropyridin-3-yl)(phenyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₉ClFNO
Storage Temperature: Ambient
CAS Number: 3037705-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR