3,5-Dichloro-4-(difluoromethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

464365-5G 464365-25G 464365-1G 464365-500MG
77633-228EA 712 USD
77633-228 77655-898 77642-986 77646-960
3,5-Dichloro-4-(difluoromethyl)pyridine ≥97%
3,5-Dichloro-4-(difluoromethyl)pyridine

Precaución: For Research Use Only

Formula: C₆H₃Cl₂F₂N
MW: 198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22124922
CAS Number: 1374659-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR