3,4-Difluoro-2-methoxybenzonitrile ≥95%

Supplier: AOBChem USA

Warning

46312-5G 46312-1G 46312-25G 46312-500MG
77633-224EA 627 USD
77633-224 77638-956 77659-472 77650-964
3,4-Difluoro-2-methoxybenzonitrile ≥95%
3,4-Difluoro-2-methoxybenzonitrile

Precaución: For Research Use Only

Formula: C₈H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD03428594
CAS Number: 886496-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR