2',4'-Dichloro-6'-methylacetanilide ≥95%

Supplier: AOBChem USA

Warning

46244-5G 46244-25G 46244-1G 46244-500MG
77635-042EA 354 USD
77635-042 77655-888 77642-984 77646-956
2',4'-Dichloro-6'-methylacetanilide ≥95%
2',4'-Dichloro-6'-methylacetanilide

Precaución: For Research Use Only

Formula: C₉H₉Cl₂NO
MW: 218.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00833408
CAS Number: 61655-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR