3-Bromo-2,4-dichoroquinoline ≥95%

Supplier: AOBChem USA

Warning

462416-5G 462416-25G 462416-1G 462416-500MG
77635-040EA 602 USD
77635-040 77659-470 77642-982 77650-962
3-Bromo-2,4-dichoroquinoline ≥95%
3-Bromo-2,4-dichoroquinoline

Precaución: For Research Use Only

Formula: C₉H₄BrCl₂N
MW: 276.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17012193
CAS Number: 109069-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR