8-Chloronaphthalene-1-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

461991-5G 461991-1G 461991-25G 461991-500MG
77633-220EA 762 USD
77633-220 77638-944 77659-466 77650-952
8-Chloronaphthalene-1-carbaldehyde ≥97%
8-Chloronaphthalene-1-carbaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD17012500
CAS Number: 129994-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR