1-(2-Aminobenzo[d]thiazol-6-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

461986-5G 461986-25G
77635-038EA 70 USD
77635-038 77655-880
1-(2-Aminobenzo[d]thiazol-6-yl)ethanone ≥95%
1-(2-Aminobenzo[d]thiazol-6-yl)ethanone

Precaución: For Research Use Only

Formula: 192.2376 g/mol
MW: C₉H₈N₂OS
Storage Temperature: Ambient
CAS Number: 21222-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR