6-Bromo-4H-1,3-benzodioxine ≥97%

Supplier: AOBChem USA

Warning

461980-5G 461980-25G
77635-036EA 125 USD
77635-036 77655-878
6-Bromo-4H-1,3-benzodioxine ≥97%
6-Bromo-4H-1,3-benzodioxine

Precaución: For Research Use Only

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10703544
CAS Number: 90050-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR