5-Ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46139-5G 46139-25G 46139-1G
77635-034EA 217 USD
77635-034 77655-876 77642-976
5-Ethoxy-2-fluorobenzaldehyde ≥97%
5-Ethoxy-2-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD24686049
CAS Number: 891842-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR