Phenyl(6-phenylpyridin-3-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

460636-5G 460636-25G 460636-1G 460636-500MG
77629-502EA 1484.23 USD
77629-502 77635-866 77638-938 77650-942
Phenyl(6-phenylpyridin-3-yl)methanone ≥97%
Phenyl(6-phenylpyridin-3-yl)methanone

Precaución: For Research Use Only

Formula: C₁₈H₁₃NO
Storage Temperature: Ambient
CAS Number: 30091-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR