1-Bromo-4-isopropyl-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

460538-5G 460538-1G 460538-500MG
77659-454EA 2513 USD
77659-454 77642-964 77650-940
1-Bromo-4-isopropyl-2-methylbenzene ≥95%
1-Bromo-4-isopropyl-2-methylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃Br
Storage Temperature: Ambient
CAS Number: 92243-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR