2-Chloro-3-methoxy-4-pyridinecarboxamide ≥97%

Supplier: AOBChem USA

Warning

460102-1G 460102-500MG 460102-5G
77638-936EA 743 USD
77638-936 77652-850 77659-452
2-Chloro-3-methoxy-4-pyridinecarboxamide ≥97%
2-Chloro-3-methoxy-4-pyridinecarboxamide

Precaución: For Research Use Only

Formula: 186.6 g/mol
MW: C₇H₇ClN₂O₂
Storage Temperature: Ambient
CAS Number: 1935682-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR