2-Chloro-3-methoxyisonicotinonitrile ≥97%

Supplier: AOBChem USA

Warning

460101-5G 460101-25G 460101-1G 460101-500MG
77633-214EA 727 USD
77633-214 77655-868 77642-960 77646-944
2-Chloro-3-methoxyisonicotinonitrile ≥97%
2-Chloro-3-methoxyisonicotinonitrile

Precaución: For Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257761
CAS Number: 1256811-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR