2,2,2-Trifluoro-1-(4-(pyridin-2-yl)phenyl)ethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
459568-5G
459568-1G
459568-25G
459568-500MG
77629-490EA
1484.23
USD
77629-490
77638-930
77655-864
77646-940
2,2,2-Trifluoro-1-(4-(pyridin-2-yl)phenyl)ethanone ≥95%
2,2,2-Trifluoro-1-(4-(pyridin-2-yl)phenyl)ethanone
Precaución: For Research Use Only
Formula:
251.2039 g/mol MW: C₁₃H₈F₃NO Storage Temperature: Ambient |
CAS Number:
2668238-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...