6-Bromo-2,3-dihydro-2-methyl-2-phenyl-4(1H)-quinazolinone ≥95%

Supplier: AOBChem USA

Warning

459518-5G 459518-25G 459518-1G 459518-500MG
77629-488EA 1484.23 USD
77629-488 77635-862 77638-928 77650-934
6-Bromo-2,3-dihydro-2-methyl-2-phenyl-4(1H)-quinazolinone ≥95%
6-Bromo-2,3-dihydro-2-methyl-2-phenyl-4(1H)-quinazolinone

Precaución: For Research Use Only

Formula: C₁₅H₁₃BrN₂O
Storage Temperature: Ambient
CAS Number: 1246861-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR