4-Ethoxy-1-iodo-2-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

458773-5G 458773-25G 458773-1G 458773-500MG
77629-486EA 1484.23 USD
77629-486 77635-860 77638-926 77650-932
4-Ethoxy-1-iodo-2-(trifluoromethyl)benzene ≥95%
4-Ethoxy-1-iodo-2-(trifluoromethyl)benzene

Precaución: For Research Use Only

Formula: 316.059 g/mol
MW: C₉H₈F₃IO
Storage Temperature: Ambient
CAS Number: 2383986-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR